Xanthene 98%

BLDPBD156620-5GEA 21 EUR
BLDPBD156620-5G BLDPBD156620-25G BLDPBD156620-100G BLDPBD156620-10G
Xanthene 98%
Xanthene

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₃H₁₀O
MW: 182,22 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005055
CAS-nummer: 92-83-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR