1-Methylpyrrole-3-carboxylic acid 95%

BLDPBD157883-250MGEA 38 EUR
BLDPBD157883-250MG BLDPBD157883-100MG BLDPBD157883-1G BLDPBD157883-5G BLDPBD157883-25G
1-Methylpyrrole-3-carboxylic acid 95%
1-Methylpyrrole-3-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C₆H₇NO₂
MW: 125,1300 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD11111733
CAS-nummer: 36929-61-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR