3-Chloro-2-methoxybenzaldehyde 97%

BLDPBD158470-1GEA 25 EUR
BLDPBD158470-1G BLDPBD158470-25G BLDPBD158470-10G BLDPBD158470-250MG BLDPBD158470-5G
3-Chloro-2-methoxybenzaldehyde 97%
3-Chloro-2-methoxybenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formule: C₈H₇ClO₂
MW: 170,5930 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09999135
CAS-nummer: 223778-54-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR