4-Amino-5-chloropyrimidine 95%

BLDPBD158492-5GEA 489 EUR
BLDPBD158492-5G BLDPBD158492-100MG BLDPBD158492-250MG BLDPBD158492-1G
4-Amino-5-chloropyrimidine 95%
4-Amino-5-chloropyrimidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₄H₄ClN₃
MW: 129,5500 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00233964
CAS-nummer: 101257-82-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR