4-Bromo-6-(trifluoromethyl)pyrimidine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD158500-250MGEA 33 EUR
BLDPBD158500-250MG BLDPBD158500-5G BLDPBD158500-100MG BLDPBD158500-1G
4-Bromo-6-(trifluoromethyl)pyrimidine 97%
4-Bromo-6-(trifluoromethyl)pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formule: C₅H₂BrF₃N₂
MW: 226,9800 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD10574962
CAS-nummer: 785777-89-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR