(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%

BLDPBD1586-1GEA 7 EUR
BLDPBD1586-1G BLDPBD1586-100G BLDPBD1586-25G BLDPBD1586-5G BLDPBD1586-500G BLDPBD1586-10G BLDPBD1586-0.025G
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₂₃H₂₅NO₆
MW: 411,5 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00077050
CAS-nummer: 112883-39-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR