3-Chloro-5-nitropyridin-4-amine 97%

BLDPBD158713-100MGEA 30 EUR
BLDPBD158713-100MG BLDPBD158713-250MG BLDPBD158713-1G BLDPBD158713-5G
3-Chloro-5-nitropyridin-4-amine 97%
3-Chloro-5-nitropyridin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C5H4ClN3O2
MW: 173,5572 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD14706168
CAS-nummer: 89284-28-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR