6-Bromo-5-chloroindoline-2,3-dione 97%

BLDPBD158842-1GEA 309 EUR
BLDPBD158842-1G BLDPBD158842-250MG BLDPBD158842-100MG
6-Bromo-5-chloroindoline-2,3-dione 97%
6-Bromo-5-chloroindoline-2,3-dione

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₃BrClNO₂
MW: 260,4700 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD10566349
CAS-nummer: 192799-05-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR