Ethyl-2,4-dichloronicotinate 98%

BLDPBD158890-1GEA 72 EUR
BLDPBD158890-1G BLDPBD158890-10G BLDPBD158890-250MG BLDPBD158890-5G
Ethyl-2,4-dichloronicotinate 98%
Ethyl-2,4-dichloronicotinate

Storage condition: Inert atmosphere, 2-8°C

Formule: C₈H₇Cl₂NO₂
MW: 220,05 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD12407388
CAS-nummer: 62022-04-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR