5-Bromo-4-methylpyridine-2,3-diamine 97%

BLDPBD159141-5GEA 184 EUR
BLDPBD159141-5G BLDPBD159141-25G BLDPBD159141-1G BLDPBD159141-250MG
5-Bromo-4-methylpyridine-2,3-diamine 97%
5-Bromo-4-methylpyridine-2,3-diamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₆H₈BrN₃
MW: 202,0518 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD08062974
CAS-nummer: 41230-93-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR