1-(5-Chloro-1H-indazol-1-yl)ethanone 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD159327-250MGEA 14 EUR
BLDPBD159327-250MG BLDPBD159327-1G
1-(5-Chloro-1H-indazol-1-yl)ethanone 95%
1-(5-Chloro-1H-indazol-1-yl)ethanone

Storage condition: Sealed in dry, 2-8°C

Formule: C₉H₇ClN₂O
MW: 194,6177 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD11109546
CAS-nummer: 98083-43-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR