1-(Pyrimidin-2-yl)piperidin-4-ol 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD159471-1GEA 41 EUR
BLDPBD159471-1G BLDPBD159471-10G BLDPBD159471-5G BLDPBD159471-250MG
1-(Pyrimidin-2-yl)piperidin-4-ol 95%
1-(Pyrimidin-2-yl)piperidin-4-ol

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₁₃N₃O
MW: 179,2200 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD05864742
CAS-nummer: 893755-98-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR