2-Bromo-4,5-difluorobenzonitrile 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD161213-25GEA 109 EUR
BLDPBD161213-25G BLDPBD161213-5G BLDPBD161213-1G BLDPBD161213-100G BLDPBD161213-10G
2-Bromo-4,5-difluorobenzonitrile 97%
2-Bromo-4,5-difluorobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₂BrF₂N
MW: 218,0000 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09864661
CAS-nummer: 64695-82-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR