5-Chloro-4-fluoro-2-nitrophenol 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD161751-250MGEA 16 EUR
BLDPBD161751-250MG BLDPBD161751-5G BLDPBD161751-25G BLDPBD161751-1G
5-Chloro-4-fluoro-2-nitrophenol 98%
5-Chloro-4-fluoro-2-nitrophenol

Storage condition: Sealed in dry, Room Temperature

Formule: C6H3ClFNO3
MW: 191,5400 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD18072578
CAS-nummer: 65001-79-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR