2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD162915-250MGEA 29 EUR
BLDPBD162915-250MG BLDPBD162915-10G BLDPBD162915-5G BLDPBD162915-100MG BLDPBD162915-1G BLDPBD162915-25G
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine 97%
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formule: C₇H₆Cl₂N₂
MW: 189,0419 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09750204
CAS-nummer: 5466-43-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR