tert-Butyl piperazin-1-ylcarbamate 97%

BLDPBD163732-5GEA 643 EUR
BLDPBD163732-5G BLDPBD163732-250MG BLDPBD163732-1G BLDPBD163732-100MG BLDPBD163732-10G
tert-Butyl piperazin-1-ylcarbamate 97%
tert-Butyl piperazin-1-ylcarbamate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₉H₁₉N₃O₂
MW: 201,2661 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD11840387
CAS-nummer: 147081-80-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR