6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine 97%

BLDPBD163857-25GEA 105 EUR
BLDPBD163857-25G BLDPBD163857-1G BLDPBD163857-10G BLDPBD163857-5G BLDPBD163857-250MG BLDPBD163857-100G
6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine 97%
6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₆H₅BrN₄
MW: 213,0400 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD11656619
CAS-nummer: 947248-68-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR