2-Chloro-6-fluoroanisole 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD164101-25GEA 39 EUR
BLDPBD164101-25G BLDPBD164101-10G BLDPBD164101-5G
2-Chloro-6-fluoroanisole 97%
2-Chloro-6-fluoroanisole
Formule: C₇H₆ClFO
MW: 160,58 g/mol
MDL-nummer: MFCD01631573
CAS-nummer: 53145-38-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR