6-Bromo-o-toluonitrile 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD164350-5GEA 133 EUR
BLDPBD164350-5G BLDPBD164350-1G BLDPBD164350-25G BLDPBD164350-100MG BLDPBD164350-10G BLDPBD164350-100G BLDPBD164350-250MG
6-Bromo-o-toluonitrile 98%
6-Bromo-o-toluonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₆BrN
MW: 196,0439 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09834775
CAS-nummer: 77532-78-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR