Benzyl-3-oxoazetidine-1-carboxylate 97%

BLDPBD164977-25GEA 69 EUR
BLDPBD164977-25G BLDPBD164977-500G BLDPBD164977-1G BLDPBD164977-100G BLDPBD164977-10G BLDPBD164977-5G BLDPBD164977-0.5G
Benzyl-3-oxoazetidine-1-carboxylate 97%
Benzyl-3-oxoazetidine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₁H₁₁NO₃
MW: 205,2100 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09954923
CAS-nummer: 105258-93-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR