4-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine 96%

Leverancier: BLD PHARMATECH GMBH

BLDPBD165424-10GEA 608 EUR
BLDPBD165424-10G BLDPBD165424-250MG BLDPBD165424-5G BLDPBD165424-1G
4-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine 96%
4-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₄H₁₁BrN₂O₂S
MW: 351,2200 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD10575009
CAS-nummer: 348640-07-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR