1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester 98%

BLDPBD165599-1GEA 92 EUR
BLDPBD165599-1G BLDPBD165599-250MG
1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester 98%
1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formule: C₁₅H₂₆BN₃O₃
MW: 307,1962 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD08062342
CAS-nummer: 864754-18-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR