3-Methylazetidine hydrochloride 95%

BLDPBD165759-250MGEA 21 EUR
BLDPBD165759-250MG
3-Methylazetidine hydrochloride 95%
3-Methylazetidine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formule: C4H10ClN
MW: 107,5800 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09743448
CAS-nummer: 935669-28-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR