7-Bromo[1,2,4]triazolo[1,5-a]pyridine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD165779-10GEA 624 EUR
BLDPBD165779-10G BLDPBD165779-1G BLDPBD165779-25G BLDPBD165779-100MG BLDPBD165779-250MG BLDPBD165779-5G
7-Bromo[1,2,4]triazolo[1,5-a]pyridine 97%
7-Bromo[1,2,4]triazolo[1,5-a]pyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₄BrN₃
MW: 198,0201 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD10699200
CAS-nummer: 1053655-66-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR