4-Butyl-4'-cyanobiphenyl 98%

BLDPBD16657-1GEA 10 EUR
BLDPBD16657-1G BLDPBD16657-5G BLDPBD16657-25G BLDPBD16657-10G
4-Butyl-4'-cyanobiphenyl 98%
4-Butyl-4'-cyanobiphenyl

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₇H₁₇N
MW: 235,33 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00506009
CAS-nummer: 52709-83-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR