3-Bromo-6-chloropicolinic acid 97%

BLDPBD167406-5GEA 33 EUR
BLDPBD167406-5G BLDPBD167406-250MG BLDPBD167406-10G BLDPBD167406-25G BLDPBD167406-1G BLDPBD167406-100G
3-Bromo-6-chloropicolinic acid 97%
3-Bromo-6-chloropicolinic acid

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₃BrClNO₂
MW: 236,4505 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09258777
CAS-nummer: 929000-66-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR