2-Methoxy-4-methylbenzaldehyde 97%

BLDPBD167585-25GEA 404 EUR
BLDPBD167585-25G BLDPBD167585-250MG BLDPBD167585-0.025 BLDPBD167585-5G BLDPBD167585-1G
2-Methoxy-4-methylbenzaldehyde 97%
2-Methoxy-4-methylbenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formule: C₉H₁₀O₂
MW: 150,1700 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD06248007
CAS-nummer: 57415-35-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR