5-Bromo-2-cyano-3-methylpyridine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD167694-100GEA 263 EUR
BLDPBD167694-100G BLDPBD167694-250MG BLDPBD167694-25G BLDPBD167694-500G BLDPBD167694-10G BLDPBD167694-5G BLDPBD167694-1G
5-Bromo-2-cyano-3-methylpyridine 98%
5-Bromo-2-cyano-3-methylpyridine

Storage condition: Inert atmosphere, Room Temperature

Formule: C₇H₅BrN₂
MW: 197,0320 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD11975788
CAS-nummer: 156072-86-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR