Methyl-1-methyl-1H-indazole-4-carboxylate 95%

BLDPBD167764-250MGEA 26 EUR
BLDPBD167764-250MG BLDPBD167764-1G BLDPBD167764-5G BLDPBD167764-100MG
Methyl-1-methyl-1H-indazole-4-carboxylate 95%
Methyl-1-methyl-1H-indazole-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₀H₁₀N₂O₂
MW: 190,1986 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD11109402
CAS-nummer: 1071428-42-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR