6-Chloro-3-pyridazinecarboxaldehyde 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD167929-250MGEA 117 EUR
BLDPBD167929-250MG BLDPBD167929-1G BLDPBD167929-100MG
6-Chloro-3-pyridazinecarboxaldehyde 95%
6-Chloro-3-pyridazinecarboxaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formule: C₅H₃ClN₂O
MW: 142,5431 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD10698041
CAS-nummer: 303085-53-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR