5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine 98%

BLDPBD168813-250MGEA 7 EUR
BLDPBD168813-250MG BLDPBD168813-5G BLDPBD168813-25G BLDPBD168813-1G BLDPBD168813-100MG BLDPBD168813-10G BLDPBD168813-0.025
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine 98%
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₆H₃BrF₃NO
MW: 242,00 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD12546486
CAS-nummer: 76041-79-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR