2-(2,6-Dibromophenyl)acetic acid 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD168834-10GEA 526 EUR
BLDPBD168834-10G BLDPBD168834-250MG BLDPBD168834-25G BLDPBD168834-1G BLDPBD168834-5G
2-(2,6-Dibromophenyl)acetic acid 97%
2-(2,6-Dibromophenyl)acetic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₆Br₂O₂
MW: 293,94 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09832981
CAS-nummer: 901310-02-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR