(5-Chloro-2-fluorophenyl)acetic acid 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD169672-25GEA 248 EUR
BLDPBD169672-25G BLDPBD169672-5G BLDPBD169672-10G BLDPBD169672-100G BLDPBD169672-1G
(5-Chloro-2-fluorophenyl)acetic acid 97%
(5-Chloro-2-fluorophenyl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formule: (C₆H₃ClF)CH₂COOH
MW: 188,59 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01631439
CAS-nummer: 261762-97-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR