(S)-3-(Boc-amino)-3-phenylpropionic acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD17344-250MGEA 32 EUR
BLDPBD17344-250MG BLDPBD17344-1G BLDPBD17344-5G
(S)-3-(Boc-amino)-3-phenylpropionic acid 98%
(S)-3-(Boc-amino)-3-phenylpropionic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₄H₁₉NO₄
MW: 265,31 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD01860892
CAS-nummer: 103365-47-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR