1-Methyl-4-nitro-1H-pyrazole 98%

BLDPBD174190-100GEA 224 EUR
BLDPBD174190-100G BLDPBD174190-1G BLDPBD174190-10G BLDPBD174190-5G BLDPBD174190-25G
1-Methyl-4-nitro-1H-pyrazole 98%
1-Methyl-4-nitro-1H-pyrazole

Storage condition: Sealed in dry, Room Temperature

Formule: C₄H₅N₃O₂
MW: 127,10 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00464261
CAS-nummer: 3994-50-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR