4-(4-Fluorophenyl)benzonitrile 98%

BLDPBD17421-1GEA 88 EUR
BLDPBD17421-1G BLDPBD17421-250MG BLDPBD17421-5G
4-(4-Fluorophenyl)benzonitrile 98%
4-(4-Fluorophenyl)benzonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₃H₈FN
MW: 197,2100 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD05981088
CAS-nummer: 10540-31-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR