(±)-2-Methylpiperazine 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD17485-25GEA 6 EUR
BLDPBD17485-25G
(±)-2-Methylpiperazine 95%
(±)-2-Methylpiperazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₅H₁₂N₂
MW: 100,16 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005954
CAS-nummer: 109-07-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR