L-Glutamic acid diethyl ester hydrochloride 95%

BLDPBD17545-10GEA 7 EUR
BLDPBD17545-10G BLDPBD17545-100G BLDPBD17545-500G BLDPBD17545-5G BLDPBD17545-25G
L-Glutamic acid diethyl ester hydrochloride 95%
L-Glutamic acid diethyl ester hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₉H₁₈ClNO₄
MW: 239,696 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00012509
CAS-nummer: 1118-89-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR