N-Boc-4-oxo-L-proline 97%

BLDPBD1782-25GEA 79 EUR
BLDPBD1782-25G BLDPBD1782-5G BLDPBD1782-100G BLDPBD1782-250MG BLDPBD1782-500G BLDPBD1782-10G BLDPBD1782-1G
N-Boc-4-oxo-L-proline 97%
N-Boc-4-oxo-L-proline

Storage condition: Inert atmosphere, 2-8°C

Formule: C₁₀H₁₅NO₅
MW: 229,23 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD01860669
CAS-nummer: 84348-37-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR