Lamivudine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD18116-5GEA 7 EUR
BLDPBD18116-5G BLDPBD18116-100G BLDPBD18116-25G BLDPBD18116-10G
Lamivudine 98%
Lamivudine

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formule: C₈H₁₁N₃O₃S
MW: 229,2600 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD00869739
CAS-nummer: 134678-17-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR