4-Nitro-2-picoline 98%

BLDPBD18125-10GEA 56 EUR
BLDPBD18125-10G BLDPBD18125-1G BLDPBD18125-25G BLDPBD18125-5G
4-Nitro-2-picoline 98%
4-Nitro-2-picoline

Storage condition: Inert atmosphere, 2-8°C

Formule: C₆H₆N₂O₂
MW: 138,1200 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD03095077
CAS-nummer: 13508-96-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR