3-(Boc-amino)-3-(2-methylphenyl)propionic acid 95%

BLDPBD182342-100MGEA 144 EUR
BLDPBD182342-100MG BLDPBD182342-1G BLDPBD182342-250MG
3-(Boc-amino)-3-(2-methylphenyl)propionic acid 95%
3-(Boc-amino)-3-(2-methylphenyl)propionic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₅H₂₁NO₄
MW: 279,3315 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD02090696
CAS-nummer: 284493-54-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR