3-Amino-4,5-dimethylisoxazole 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD18258-1GEA 76 EUR
BLDPBD18258-1G BLDPBD18258-250MG BLDPBD18258-25G BLDPBD18258-100MG BLDPBD18258-5G
3-Amino-4,5-dimethylisoxazole 98%
3-Amino-4,5-dimethylisoxazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C5H8N2O
MW: 112,1300 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03411577
CAS-nummer: 13999-39-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR