(S)-Ethyl-N-boc-pyroglutamate 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD18378-10GEA 7 EUR
BLDPBD18378-10G BLDPBD18378-1000G BLDPBD18378-500G BLDPBD18378-25G BLDPBD18378-5G BLDPBD18378-100G
(S)-Ethyl-N-boc-pyroglutamate 97%
(S)-Ethyl-N-boc-pyroglutamate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₂H₁₉NO₅
MW: 257,3 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD07778090
CAS-nummer: 144978-12-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR