5-Bromo-2-hydroxy-4-picoline 95%

BLDPBD18739-25GEA 84 EUR
BLDPBD18739-25G BLDPBD18739-500G BLDPBD18739-5G BLDPBD18739-100G BLDPBD18739-1G BLDPBD18739-10G
5-Bromo-2-hydroxy-4-picoline 95%
5-Bromo-2-hydroxy-4-picoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₆BrNO
MW: 188,02 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03427664
CAS-nummer: 164513-38-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR