(R)-2-Amino-3-(tritylthio)propanamide 95%

BLDPBD18778-250MGEA 10 EUR
BLDPBD18778-250MG BLDPBD18778-1G BLDPBD18778-25G BLDPBD18778-5G BLDPBD18778-10G
(R)-2-Amino-3-(tritylthio)propanamide 95%
(R)-2-Amino-3-(tritylthio)propanamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₂₂H₂₂N₂OS
MW: 362,49 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD22126061
CAS-nummer: 166737-85-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR