3-(4-Fluorophenoxy)pyrrolidine hydrochloride 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD188572-250MGEA 93 EUR
BLDPBD188572-250MG BLDPBD188572-1G
3-(4-Fluorophenoxy)pyrrolidine hydrochloride 95%
3-(4-Fluorophenoxy)pyrrolidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₀H₁₃ClFNO
MW: 217,6700 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09879244
CAS-nummer: 23123-11-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR