(1-Methyl-1H-imidazol-2-yl)methanol 97%
|
|
BLDPBD18867-100GEA
111
EUR
BLDPBD18867-100G
BLDPBD18867-5G
BLDPBD18867-500G
BLDPBD18867-10G
BLDPBD18867-25G
(1-Methyl-1H-imidazol-2-yl)methanol 97%
(1-Methyl-1H-imidazol-2-yl)methanol
Storage condition: Inert atmosphere, Room Temperature
Formule:
C₅H₈N₂O MW: 112,13 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00964673 CAS-nummer: 17334-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...