Methyl-3-(cyclopentylamino)propanoate 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD192847-10GEA 492 EUR
BLDPBD192847-10G BLDPBD192847-1G BLDPBD192847-250MG BLDPBD192847-5G
Methyl-3-(cyclopentylamino)propanoate 95%
Methyl-3-(cyclopentylamino)propanoate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₉H₁₇NO₂
MW: 171,2368 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD10687312
CAS-nummer: 754125-43-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR