5-Bromo-1,3,4-thiadiazol-2-amine 98%

BLDPBD1938-5GEA 11 EUR
BLDPBD1938-5G BLDPBD1938-100G BLDPBD1938-1G BLDPBD1938-500G BLDPBD1938-0.025G BLDPBD1938-10G BLDPBD1938-25G
5-Bromo-1,3,4-thiadiazol-2-amine 98%
5-Bromo-1,3,4-thiadiazol-2-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₂H₂BrN₃S
MW: 180,02 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00464325
CAS-nummer: 37566-39-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR